Merck
US EPA Method 0030: GC Analysis of Volatiles on SPB®-624 after Collection/Desorption using VOST 100 Tube
US EPA Method 0030: GC Analysis of Volatiles on SPB®-624 after Collection/Desorption using VOST 100 Tube suitable for GC, application for air monitoring
材料
相關產品
產品號碼
描述
訂價
Vocarb® 4000 吹扫/捕集器 I
for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT-3
VOST 堆叠采样管
35-60 mesh
CONDITIONS
sample
calibration standard: volatiles in methanol; blank: Tenax TA tube with surrogate thermally desorbed into purge & trap system
adsorbent tube
VOST 100 (20074-U)
purge trap
VOCARB 4000 (20308)
column
SPB-624, 75 m × 0.53 mm I.D., 3.0 μm film (25432)
oven
35 °C (5 min) to 200 °C at 10 °C/min, hold 2 min
detector
FID, 300 °C
carrier gas
helium, 8 mL/min
描述
法律資訊
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany