GC Analysis of Phenylethanol Enantiomers on β-DEX™ 120
材料
分析柱
產品號碼
描述
訂價
CONDITIONS
column
β-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24304)
oven
120 °C
inj. temp.
200 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec
injection
1 μL, 100:1 split
sample
0.5 mg/mL each analyte in methylene chloride
描述
法律資訊
DEX is a trademark of Sigma-Aldrich Co. LLC