重要文件
ZUFPREUN0
Milli-Q
UF Pretreatment Unit 3/4
Removes contaminants from tap water with fouling index above system specifications
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41104214
NACRES:
JA.13
暫時無法取得訂價和供貨情況
AC/DC 輸入
6 V
包裝
pkg of 1 unit
參數
22 kg operating weight (48.5 lb)
尺寸高度 × 寬度 × 直徑
129 cm (50.8 in.) × 30 cm (11.8 in.) × 25 cm (9.8 in.)
相容性
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
一般說明
This ultrafiltration (UF) pretreatment unit removes suspended particulates, microorganisms, and colloids from a tap water and is used when tap water has a high Fouling Index (7 > SDI > 12).
應用
Pretreats tap water with high fouling index before being fed to Milli-Q® CLX/HX/HX-SD/HR 7000 systems.
法律資訊
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany