Skip to Content
Merck

Skip To

33427

2,4-TDI

analytical standard

Synonym(s):

Toluene 2,4-diisocyanate, 2,4-Diisocyanatotoluene, 4-Methyl-m-phenylene diisocyanate, Tolylene 2,4-diisocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H6N2O2
CAS Number:
Molecular Weight:
174.16
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
209-544-5
Beilstein/REAXYS Number:
744602
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

bp

124-126 °C/18 mmHg (lit.), 251 °C (lit.)

mp

19.5-21.5 °C (lit.)

density

1.22 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

Cc1ccc(cc1N=C=O)N=C=O

InChI

1S/C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3

InChI key

DVKJHBMWWAPEIU-UHFFFAOYSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here
2,4-TDI is an environmental contaminant belonging to the class of diisocyanates. It is commercially used as a starting material for varnishes, paints, elastomers, polyurethane foams, wire enamels, etc.

Application

2,4-TDI may be used as an analytical reference standard for the quantification of the analyte in gas-phase and biological fluids using chromatography techniques.


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

269.6 °F - closed cup

flash_point_c

132 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters