主要文件
56034
Omnifit® 标准口径连接器
Three-way valve connector
登录查看公司和协议定价
选择尺寸
1 EA
$256.00
$256.00
请联系客服了解存货情况
获取大包装报价
选择尺寸
变更视图
1 EA
$256.00
About This Item
UNSPSC代码:
12000000
$256.00
请联系客服了解存货情况
获取大包装报价
技术
LPLC: suitable
1.5 mm
相容性
for use with 1/16 in. tubing
正在寻找类似产品? 访问 产品对比指南
一般描述
Omnifit® Standard Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The standard bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.[1]
应用
Omnifit® Standard Bore Connector was used to construct the flow-injection system for amperometric determination of tacrine.[1]
特点和优势
包括 O 形圈和 PTFE 锥。
法律信息
Omnifit is a registered trademark of Diba Industries
历史批次信息供参考:
分析证书(COA)
访问文档库
Modern Instrumental Techniques
Keith, A
Soil and Environmental Analysis, 1-700 (2003)
Flow-Injection Amperometric Determination of Tacrine based on Ion Transfer across a Water-Plasticized Polymeric Membrane Interface.
Joaquin, A. C.
Sensors, 7, 1185-1192 (2007)