主要文件
Z274240
Sigma-Aldrich
Aldrich® tube fitting kit
for tubing i.d., 1/8 in.
登录查看公司和协议定价
选择尺寸
1 KIT
$353.00
$353.00
请联系客服了解存货情况
选择尺寸
变更视图
1 KIT
$353.00
About This Item
UNSPSC代码:
41121815
NACRES:
NB.35
$353.00
请联系客服了解存货情况
特点
for tubing i.d. 1/8 in.
正在寻找类似产品? 访问 产品对比指南
一般描述
Clear, natural Nylon-66 fittings for making a variety of connections with flexible tubing. Fittings are supplied in a compartmentalized PP storage box.
Contains 165 fittings that include: tubing hose barbs, connectors, tees, branched tees, elbows, plugs, reducers, and locking sleeves.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)