26891U
26891-U
$757.00
1 of 10
26264
配备可拆卸针头的 SGE 气密注射器
Sigma-Aldrich
X100
Triton™ X-100
T8787
Supelco
24833-U
色谱柱的螺母
26256
SGE气密注射器,固定针头
26266
15S9
TERGITOL烷氧基聚乙烯氢氧基乙醇(非离子表面活性剂)™
24807-U
Supeltex M-2A Capillary Ferrule, Short Design
P5368
Phosphate buffered saline
26263
cone tip stainless steel needle (point style 1)
23-26 ga
color (dark orange)
needle type fixed tapered needlesyringe type Gas tight
pkg of 1 × 6 ea (26891-U)
SGE
42 mm
10 μL
正在寻找类似产品? 访问 产品对比指南
Analytical Chromatography
Analytical Syringes
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.