主要文件
22284
按钮阀
for use with VICI Series C and D Pressure-Lok syringes
登录查看公司和协议定价
选择尺寸
1 EA
$47.80
$47.80
预计发货时间2025年5月22日详情
获取大包装报价
选择尺寸
变更视图
1 EA
$47.80
About This Item
分類程式碼代碼:
41122001
$47.80
预计发货时间2025年5月22日详情
获取大包装报价
材料
PTFE Mininert valve
包裝
pkg of 1 ea
1 ea of
製造商/商標名
654050
VICI Precision Sampling
相容性
for use with VICI Series C and D Pressure-Lok syringes
正在寻找类似产品? 访问 产品对比指南
一般說明
This Mininert valve will attach to your syringe and provide sample storing capability. It features push-button operation and an all-PTFE body with a stainless steel miniature valve stem.
历史批次信息供参考:
分析证书(COA)