主要文件
Z377619
Sigma-Aldrich
Flow-Mesh gel and membrane support
sheet size 8 1/2 in. × 11 in., pkg of 20 sheets
登录查看公司和协议定价
选择尺寸
1 PKG
$91.30
$91.30
请联系客服了解存货情况
选择尺寸
变更视图
1 PKG
$91.30
About This Item
UNSPSC代码:
41105300
NACRES:
NB.51
$91.30
请联系客服了解存货情况
包装
pkg of 20 sheets
制造商/商品名称
Diversified Biotech FM-100
薄片尺寸
8 1/2 in. × 11 in.
一般描述
Inert perforated mesh with a high fraction of open volume, permitting free diffusion of solutions around gels. Use mesh to support gels and membranes for any solution treatment: staining/destaining, probing, incubation, etc. Supported gels and membranes will not tear or bend, and may be stacked for efficient use of time and solutions.
May be cut to size with scissors.
法律信息
Flow-Mesh is a trademark of C.B.S. Scientific Co.
历史批次信息供参考:
分析证书(COA)