主要文件
Z376809
Sigma-Aldrich
CryoTube® work station, autoclavable
pkg of 4
登录查看公司和协议定价
选择尺寸
1 PKG
$168.00
$168.00
请联系客服了解存货情况
选择尺寸
变更视图
1 PKG
$168.00
About This Item
UNSPSC代码:
41121800
NACRES:
NB.22
$168.00
请联系客服了解存货情况
物料
red
包装
pkg of 4
制造商/商品名称
Simport T315-3
尺寸
4 in. × 8 in. × 1 in.
一般描述
An autoclavable rack that holds up to 50 microcentrifuge tubes. Each place is identified with an alpha-numeric index. Unique locking system allows for the tubes to be locked into place while caps may be removed with one hand.
法律信息
CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
历史批次信息供参考:
分析证书(COA)