EP0030128796
1 of 10
Z101141
Stainless steel 304 syringe needle
Sigma-Aldrich
916757
Stainless steel dispensing tips
26274
旋转锁定注射器的 SGE 针头
19136-U
Hamilton ® RN注射器针头
T1418
Nalgene ® 离心管,橡树岭样式 3119
BAH195700000
SP Bel-Art® Tubing Connector Set
26273-U
CLS430791
Corning® 15 mL离心管
Z105058
Tubing fitting assortment kit
Z683531
无针头 PP/PE 注射器
conical bottom colorless wellsgreen plate
non-sterile
skirt
pkg of 300 ea
Eppendorf® 0030128796
PCR: suitable
96 wells
150 μm