Merck
Z677876
Finntip® Flex pipette tips
Finntip Flex 200, sterile
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
材料
colorless tip
yellow
無菌
sterile
特點
barrier: no
包裝
pack of 960 ea (10 racks of 96 tips)
製造商/商標名
Finn 94060313
最大體積
200 μL
容積範圍
1-200 μL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
其他說明
Browse all pipette tips
法律資訊
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries