Merck
Z190519
Sigma-Aldrich
液体闪烁瓶(带螺纹瓶盖)
urethane foam-filled high-density polyethylene cap, metal foil liner, pkg of 500 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121806
NACRES:
NB.11
材料
clear glass vial
metal foil liner
urethane foam-filled high-density polyethylene cap
特點
closure type screw top vial
closure type white urea cap
包裝
pkg of 500 ea
尺寸
28 mm × 61 mm
容積
20 mL
接頭
thread for 22-400
正在寻找类似产品? 访问 产品对比指南
一般說明
Autoclavable, easy to maintain. A vial is used to store or as a closure type,in order to prevent contamination.
可靠的 Wheaton "180" 牌低钾玻璃。白色 22mm 瓶盖具有可书写的顶部。带盖样品瓶在 100 只装托盘中单格包装。