主要文件
CLS70792N
Pyrex® serological pipettes, disposable, non-sterile, unplugged
volume 2.0 mL
别名:
pipet
登录查看公司和协议定价
选择尺寸
700 EA
$329.00
$329.00
请联系客服了解存货情况
选择尺寸
变更视图
700 EA
$329.00
About This Item
UNSPSC代码:
41121500
NACRES:
NB.04
$329.00
请联系客服了解存货情况
物料
borosilicate glass
无菌性
non-sterile
特点
DNase free: no
RNase free: no
graduations
subdivision
包装
case of 700 ea
制造商/商品名称
Corning 7079-2N
体积测量范围
2.0 mL
容量
2.0 mL
吸头尺寸
, Narrow
正在寻找类似产品? 访问 产品对比指南
特点和优势
  • Disposable PYREX® glass pipettes are calibrated "to deliver" (TD) with blow-out
  • Long, slender, tapered tips of pipettes make pipetting faster and easier
  • All pipettes have negative graduations and fire polished tips for burr-free, even flow rates
  • Nonsterile, unplugged and bulk packed.
法律信息
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)