Merck
Establishment of optimal conditions for long-term culture of erythrocytic stages of Theileria uilenbergi.
American journal of veterinary research (2006-11-03)
Joana P G Miranda, Elisabete M Nascimento, Helder J Cruz, Hong Yin, Erich Zweygarth, Abel G Oliva
ABSTRAKT
To establish optimal conditions for long-term culture of the erythrocytic stage of Theileria uilenbergi. Red blood cells from 3 splenectomized sheep experimentally infected with a blood stabilate of T uilenbergi. Cultures of T uilenbergi were initiated by use of blood from experimentally infected sheep collected when parasites were detected in Giemsa-stained thin blood smears. Different culture conditions were tested to optimize in vitro growth of the organisms. Subcultures were performed at a ratio of 1:2, 1:4, and 1:8 when the percentage of parasitized erythrocytes (PPE) was at least 1% or when the initial PPE was doubled. The optimal culture medium was HL-1 medium (a complete chemically defined medium) supplemented with 20% sheep serum and 0.75% chemically defined lipids. Optimal culture conditions included incubation in a humidified 2% O(2), 5% CO(2), and 93% N(2) atmosphere at 37 degrees C. Cultures of the merozoite stage of the parasite were continuously propagated in vitro for > 1 year. The PPE reached values of up to 3%. Optimization of culture conditions to reach a high PPE seems worthwhile. The continuous propagation of T uilenbergi in culture allows the production of parasite material without infecting animals and provides a continuous laboratory source of parasites for further studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sera from sheep
Sigma-Aldrich
Sheep Serum, USA origin, sterile-filtered, suitable for cell culture