Merck
Inhibition of isolated Mycobacterium tuberculosis fatty acid synthase I by pyrazinamide analogs.
Antimicrobial agents and chemotherapy (2007-05-09)
Silvana C Ngo, Oren Zimhony, Woo Jin Chung, Halimah Sayahi, William R Jacobs, John T Welch
ABSTRAKT
An analog of pyrazinamide (PZA), 5-chloropyrazinamide (5-Cl-PZA), has previously been shown to inhibit mycobacterial fatty acid synthase I (FASI). FASI has been purified from a recombinant strain of M. smegmatis (M. smegmatis Deltafas1 attB::M. tuberculosis fas1). Following purification, FASI activity and inhibition were assessed spectrophotometrically by monitoring NADPH oxidation. The observed inhibition was both concentration and structure dependent, being affected by both substitution at the 5 position of the pyrazine nucleus and the nature of the ester or N-alkyl group. Under the conditions studied, both 5-Cl-PZA and PZA exhibited concentration and substrate dependence consistent with competitive inhibition of FASI with K(i)s of 55 to 59 microM and 2,567 to 2,627 microM, respectively. The results were validated utilizing a radiolabeled fatty acid synthesis assay. This assay showed that FASI was inhibited by PZA and pyrazinoic acid as well as by a series of PZA analogs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzamide, 99%
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Pyrazinecarboxylic acid, 99%
Sigma-Aldrich
Benzamide, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrazinecarboxamide