Merck
A novel transcriptional coactivator, p52, functionally interacts with the essential splicing factor ASF/SF2.
Molecular cell (1999-01-14)
H Ge, Y Si, A P Wolffe
ABSTRAKT
Increasing evidence suggests that pre-mRNA splicing can take place cotranscriptionally in vivo. However, insight into how these two processes are linked has been lacking. Here, we describe that a novel transcriptional coactivator, p52, interacts not only with transcriptional activators and general transcription factors to enhance activated transcription but also with the essential splicing factor ASF/SF2 both in vitro and in vivo to modulate ASF/SF2-mediated pre-mRNA splicing. Furthermore, immunofluorescence studies indicate that the majority of endogenous p52 is colocalized with ASF/SF2 in the nucleoplasm of HeLa cells. Together, these observations suggest that, in addition to functioning as a transcriptional coactivator, p52 may also act as an adaptor to coordinate pre-mRNA splicing and transcriptional activation of class II genes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p52 human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Sigma-Aldrich
p75 human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)