Wstecz
Automatic Sanitization Module (ASM)
Products
Kategoria produktu
UV lamps (6)
Marka
Milli-Q (6)
Długość (cm)
Długość (cale)
Kompatybilność
Filtruj produkty
Showing 1-6 of 6
Porównaj
Numer produktu
Opis
Cennik
TANKASMES
Automatic Sanitization Module (ASM),
For use with RiOs
™
/Elix
®
/AFS
®
Essential systems, Prevents bacterial growth and limits the formation of biofilm inside the storage tank
Rozwiń
Wyświetl ceny
TANKS60UV
Automatic Sanitization Module (ASM),
AC/DC input 120 V, Protects stored pure (Type 2/3) water from bacterial proliferation and biofilm buildup.
Rozwiń
Wyświetl ceny
TANKASMIN
Automatic Sanitization Module (ASM),
For use with Elix
®
and Milli-Q
®
Direct systems, Limits the development of biofilm inside the storage tank
Rozwiń
Wyświetl ceny
TANKSDS2ASM
Automatic Sanitization Module (ASM),
Protects and preserves the integrity of stored pure water against bacterial proliferation and biofilm formation
Rozwiń
Wyświetl ceny
TANKS50UV
Automatic Sanitization Module (ASM),
AC/DC input 230 V, Protects stored pure (Type 2/3) water from bacterial proliferation and biofilm buildup.
Rozwiń
Wyświetl ceny
TANKASMUV
Automatic Sanitization Module (ASM),
For use with Large Systems, Inhibits the biofilm build-up inside the SDS-200 and 350-liter storage tanks