Przejdź do
57272
Visiprep™ Large Volume Sampler
for use with 12, 20, or 60mL SPE tubes (includes 3 adapters)
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1450,00 zł
Informacje o tej pozycji
UNSPSC Code:
12352200
NACRES:
NB.21
eCl@ss:
32060107
1450,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówieniePoproś o próbkę
Pomoc techniczna
Pozwól nam pomóc
technique(s)
solid phase extraction (SPE): suitable
Quality Segment
application(s)
food and beverages
compatibility
for use with 12, 20, or 60mL SPE tubes (includes 3 adapters), technique used for transfers 3 large volume samples
General description
For continuous "hands off" direct transfer of multiple liquid samples
A Visiprep large volume sampler enables you to transfer low viscosity samples directly from any sample container to conventional solid phase extraction tubes on a Visiprep SPE vacuum manifold.
Two samplers are available. One sampler has three tube adapters compatible with 12mL, 20mL, or 60mL polypropylene SPE tubes. Simultaneous extraction of 12 samples on a 12-port Visiprep manifold requires four of these samplers. The other sampler has four tube adapters for 3mL or 6mL SPE tubes. To simultaneously extract 12 samples, three samplers are required.
The samplers consist of 1/8-inch PTFE tubing with a screw fitting at one end and a stainless steel weight at the other end. To use the sampler, feed the weighted end into a sample container until it touches the bottom. Insert the tube adapters into the conditioned SPE tubes on your Visiprep manifold. Turn on the vacuum to the Visiprep manifold and open the manifold flow control valves, and the samples will be delivered to the SPE tubes. The flow rate of sample through each tube can be independently controlled by using the flow control valves. Both the screw fittings and the PTFE tubing on the samplers are colorcoded for easy sample identification.
Also fits Preppy SPE Vacuum Manifold.
Note: The Visiprep Large Volume Sampler can only be used with polypropylene SPE tubes. You must equip alternate manifold valves with long stem flow control knobs to accommodate 12 mL, 20 mL, or 60 mL SPE tubes.
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
572755727857277
description
for use with 12, 20, or 60mL SPE tubes (includes 3 adapters)
description
for use with 3 or 6 mL SPE tubes (includes 4 adapters)
description
weight fitting for 1/8 in. O.D. tubing, pk of 4
description
color-coded, pk of 4
application(s)
food and beverages
application(s)
PFAS testing
food and beverages
application(s)
food and beverages
application(s)
food and beverages
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
compatibility
technique used for transfers 3 large volume samples, for use with 12, 20, or 60mL SPE tubes (includes 3 adapters)
compatibility
for use with 3 or 6 mL SPE tubes (includes 4 adapters), technique used for transfers 4 large volume samples
compatibility
-
compatibility
-
Quality Level
100
Quality Level
100
Quality Level
-
Quality Level
-
Still not finding the right product?
Explore all of our products under Visiprep™ Large Volume Sampler
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)