Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 666,00 zł |
Informacje o tej pozycji
666,00 zł
material
black phenolic cap (open-top), clear glass vial, tan PTFE/silicone septum
feature
closure type black phenolic hole cap, closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
15 mm × 45 mm
volume
4 mL
fitting
thread for 13-425
Application
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 4 mL, clear glass vial, O.D. × H 15 mm × 45 mm, tan PTFE/silicone septum | description volume 40 mL, clear glass vial, O.D. × H 29 mm × 82 mm, tan PTFE/silicone septum | description volume 15 mL, clear glass vial, O.D. × H 21 mm × 70 mm, tan PTFE/silicone septum | description volume 7 mL, amber glass vial, O.D. × H 17 mm × 60 mm, tan PTFE/silicone septum |
| fitting thread for 13-425 | fitting thread for 24-400 | fitting thread for 18-400 | fitting thread for 15-425 |
| volume 4 mL | volume 40 mL | volume 15 mL | volume 7 mL |
| feature closure type black phenolic hole cap, closure type screw top vial | feature closure type black phenolic hole cap, closure type screw top vial | feature closure type black phenolic hole cap, closure type screw top vial | feature closure type black phenolic hole cap, closure type screw top vial |
| material black phenolic cap (open-top), tan PTFE/silicone septum, clear glass vial | material black phenolic cap (open-top), clear glass vial, tan PTFE/silicone septum | material black phenolic cap (open-top), clear glass vial, tan PTFE/silicone septum | material amber glass vial, black phenolic cap (open-top), tan PTFE/silicone septum |
| O.D. × H 15 mm × 45 mm | O.D. × H 29 mm × 82 mm | O.D. × H 21 mm × 70 mm | O.D. × H 17 mm × 60 mm |
| packaging pkg of 100 ea | packaging pkg of 100 ea | packaging pkg of 100 ea | packaging pkg of 100 ea |
Still not finding the right product?
Explore all of our products under Vials, screw top with phenolic open-top cap, pre-assembled
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?