22781
Właściwości
glass
100
suitable for EPA 601
Tekmar® 14-2337-024
5 mL
air monitoring (Devices & Tubes)industrial hygiene
for use with Tekmar® 2000/3000, for use with standard aqueous analysis
Opis
Pokaż różnice
1 of 1
Supelco
Fritted Purge Sampler
22789
22749
25084
Empty Tube for Thermal Desorption
size 5 mL, Tekmar® 14-2337-024
size 25 mL, Tekmar® 14-2334-024
size 25 mL, top stem, Tekmar® 14-4007-024
O.D. × L 1/4 in. × 3 1/2 in.
industrial hygiene
-
EPA 603
glass tube
Tekmar® 14-2334-024
Tekmar® 14-4007-024
25 mL
Explore all of our products under Fritted Purge Sampler
Wybierz jedną z najnowszych wersji:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Odwiedź Bibliotekę dokumentów
Powiązane kategorie
Air Monitoring Devices & Tubes
Analytical Chemistry
Analytical Sample Preparation