Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 483,00 zł 362,25 zł |
Informacje o tej pozycji
362,25 zł
Cena katalogowa483,00 złZaoszczędź 25%Quality Segment
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Sustainability features
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
- VOST Tubes (Volatile Organic Sampling Train) are designed to meet specifications in US EPA SW-846, Method 0030.
- Each tube is individually numbered, pre-conditioned, and sealed with stainless steel SWAGELOK fittings before stored in a glass storage container.
- Each lot is tested for background and backpressure.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description empty | description for use with all TDS3 Containers | description pkg of 20 ea | description O.D. × L 1/4 in. × 3 1/2 in. |
| technique(s) active air sampling: suitable | technique(s) active air sampling: suitable | technique(s) passive air sampling: suitable | technique(s) - |
| Quality Level 100 | Quality Level - | Quality Level - | Quality Level - |
| application(s) air monitoring | application(s) air monitoring | application(s) air monitoring | application(s) industrial hygiene |
| greener alternative category | greener alternative category | greener alternative category | greener alternative category |
| greener alternative product characteristics Waste Prevention | greener alternative product characteristics Waste Prevention | greener alternative product characteristics Waste Prevention | greener alternative product characteristics Waste Prevention |
| sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability Greener Alternative Product |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?