Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 5150,00 zł 3862,50 zł |
Informacje o tej pozycji
3862,50 zł
Cena katalogowa5150,00 złZaoszczędź 25%material
bevel tip needle, glass syringe
description
Magnum Series
feature
removable needle
packaging
pkg of 1 ea
manufacturer/tradename
VICI 060042
needle L × O.D. × I.D.
2 in. × 0.035 in. × 0.023 in.
volume
500 mL
application(s)
general analytical
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 500 mL, removable needle, needle L × O.D. × I.D. 2 in. × 0.035 in. × 0.023 in. | description volume 100 mL, removable needle, needle L × O.D. × I.D. 2 in. × 0.035 in. × 0.023 in. | description volume 1000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. | description volume 500 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.25 in. × 0.029 in. × 0.012 in. |
| material bevel tip needle, glass syringe | material glass syringe, bevel tip needle | material bevel tip stainless steel needle (point style 2) | material bevel tip stainless steel needle (point style 2) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| volume 500 mL | volume 100 mL | volume 1000 μL | volume 500 μL |
| manufacturer/tradename VICI 060042 | manufacturer/tradename VICI 060040 | manufacturer/tradename VICI 030033 | manufacturer/tradename VICI 010032 |
| feature removable needle | feature removable needle | feature removable needle | feature removable needle Series A |
| needle L × O.D. × I.D. 2 in. × 0.035 in. × 0.023 in. | needle L × O.D. × I.D. 2 in. × 0.035 in. × 0.023 in. | needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. | needle L × O.D. × I.D. 2.25 in. × 0.029 in. × 0.012 in. |
Still not finding the right product?
Explore all of our products under Valco® Precision Sampling syringe, Magnum series
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?