Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
0.5-10 μL
Packaging:
pack of 1 ea
Feature:
12-channel, electronic: no
Manufacturer/tradename:
Sartorius 728220
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócfeature
12-channel, electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728220
volume
0.5-10 μL
Features and Benefits
- The click-stop system enables easy volume adjustment.
- Broad volume display for better reading.
- Color-coded for easy matching of fitting tips
- High chemical and UV-resistance materials to ensure a long lifetime of the pipette.
- Fully autoclavable and requires no disassembly.
- Easy cleaning and maintenance with only three parts to disassemble.
- Safe-cone filters provided for models >10μL.
- Εrgοnomic finger support reduces the grip force required to hold the pipette.
- Range of volume-3μL (5μL for fixed) to 10mL
- Low pipetting forces
- Wide range of adjustable single and multi-channel models and fixed single-channel models
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 12-channel, volume 0.5-10 μL | description 8-channel, volume 0.5-10 μL | description 8-channel, volume 30-300 μL | description single channel, volume 1-10 μL |
| manufacturer/tradename Sartorius 728220 | manufacturer/tradename Sartorius 728120 | manufacturer/tradename Sartorius 728140 | manufacturer/tradename Sartorius 728020 |
| volume 0.5-10 μL | volume 0.5-10 μL | volume 30-300 μL | volume 1-10 μL |
| feature 12-channel, electronic: no | feature 8-channel, electronic: no | feature electronic: no, 8-channel | feature electronic: no, single-channel |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Sartorius Proline® Plus multichannel mechanical pippettor
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?