Kluczowe dokumenty
Z689130
Jenco stereo zoom microscope, GL6 series
trinocular, 10X - 65X magnification, dual halogen, AC/DC input 220 V AC, UK plug
About This Item
Kod UNSPSC:
41100000
NACRES:
MA.44
producent / nazwa handlowa
Jenco
Opis ogólny
The GL Series is par-focal and its continuous zoom is available with detentes at 0.5X increments for precise measurements. With superb resolution the GL Series is ideal for semi-conductor, high-tech industries, medical device assembly and life science research.
The comfort design allows for long duration tasks making them less fatiguing with accessible controls. Bonders and probers provide a compact design, and a long working distance, and the microscopes are fully integrated with camera and software systems.
All models include: a pair of 10X eye pieces, black/white reversible stage plate, dust cover, power cord and instruction manual.
  • Wide interpupillary adjustment (45 - 75mm) and diopters built-in to the eyepieces
  • Side mounted zoom controls and low positioned focus control with tension adjustment
  • The clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design.
  • A 100 mm working distance
  • Large zoom range gives accurate reproduction of the sample up to 260X
Informacje prawne
Jenco is a trademark of Jenco International, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)