Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
6 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 185,00 zł |
Informacje o tej pozycji
185,00 zł
material
flat bottom, polypropylene , wide-neck
joint
joint ST/NS45/40, joint neck GL52
sterility
non-sterile
feature
closure type Screw cap, with cap
packaging
case of 6
manufacturer/tradename
BRAND 93175
O.D. × H
85 mm × 140 mm
capacity
250 mL
General description
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 250 mL | description capacity 1,000 mL | description capacity 100 mL | description capacity 500 mL |
| manufacturer/tradename BRAND 93175 | manufacturer/tradename BRAND 93185 | manufacturer/tradename BRAND 93172 | manufacturer/tradename BRAND 93180 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| capacity 250 mL | capacity 1,000 mL | capacity 100 mL | capacity 500 mL |
| material polypropylene , flat bottom, wide-neck | material polypropylene , flat bottom, wide-neck | material polypropylene , wide-neck , flat bottom | material flat bottom, wide-neck , polypropylene |
| feature with cap, closure type Screw cap | feature closure type Screw cap, with cap | feature closure type Screw cap, with cap | feature closure type Screw cap, with cap |
| joint joint ST/NS45/40, joint neck GL52 | joint joint ST/NS45/40, joint neck GL52 | joint joint ST/NS34/35, joint neckGL 40 | joint joint neck GL52, joint ST/NS45/40 |
Still not finding the right product?
Explore all of our products under BRAND® Erlenmeyer flask, wide-neck, screw cap, PP
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?