Przejdź do
Z331643
BRAND® Erlenmeyer flask, wide-neck, screw cap, PP
capacity 500 mL
Synonim(y):
conical flasks
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
500 mL
Material:
flat bottom, polypropylene , wide-neck
Sterility:
non-sterile
Feature:
closure type Screw cap, with cap
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
flat bottom, polypropylene , wide-neck
joint
joint ST/NS45/40, joint neck GL52
sterility
non-sterile
feature
closure type Screw cap, with cap
packaging
case of 6
manufacturer/tradename
BRAND 93180
O.D. × H
108 mm × 180 mm
capacity
500 mL
General description
High clarity. With graduations. Threaded neck with standard ground socket.
Other Notes
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z331619Z331627Z331635
description
capacity 500 mL
description
capacity 50 mL
description
capacity 100 mL
description
capacity 250 mL
capacity
500 mL
capacity
50 mL
capacity
100 mL
capacity
250 mL
manufacturer/tradename
BRAND 93180
manufacturer/tradename
BRAND 93170
manufacturer/tradename
BRAND 93172
manufacturer/tradename
BRAND 93175
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
material
polypropylene , flat bottom, wide-neck
material
polypropylene , wide-neck , flat bottom
material
flat bottom, polypropylene , wide-neck
material
flat bottom, polypropylene , wide-neck
feature
with cap, closure type Screw cap
feature
with cap, closure type Screw cap
feature
with cap, closure type Screw cap
feature
with cap, closure type Screw cap
joint
joint neck GL52, joint ST/NS45/40
joint
joint neckGL 40, joint ST/NS34/35
joint
joint neckGL 40, joint ST/NS34/35
joint
joint ST/NS45/40, joint neck GL52
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)