Wybierz wielkość
1600,00 zł
1600,00 zł
Informacje o tej pozycji
Przejdź do
material
clear bottom, polystyrene , round black/clear bottom
sterility
sterile
feature
lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface)
packaging
pkg of 5 ea
manufacturer/tradename
Corning 4516
size
384 wells
well volume
90 μL
wells
384 wells
working volume
20-80 μL
suitability
suitable for (cell cuture)
binding type
(ultra-low attachment)
1 of 4
Ta pozycja | |||
|---|---|---|---|
| material clear bottom, round black/clear bottom , polystyrene | material polystyrene , round black/clear bottom | material clear bottom, polystyrene , round black/clear bottom | material clear bottom, polystyrene , round black/clear bottom |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| feature lid | feature ULA (Ultra-Low Attachment surface), Lid Included, Recommended Medium Well Volume, Recommended Working Volume, Specific plates for 3D cell culture | feature lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface) | feature lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface) |
| packaging pkg of 5 ea | packaging - | packaging pkg of 5 ea | packaging pkg of 10 ea, pkg of 50 ea |
| manufacturer/tradename Corning 4516 | manufacturer/tradename Corning 4527 | manufacturer/tradename Corning 4515 | manufacturer/tradename Corning 4520 |
| well volume 90 μL | well volume (14 mL) | well volume 300 μL | well volume 300 μL |
General description
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel proprietary well geometry aids formation of spheroids in center of well
- Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
- Uniform, single spheroid formation across all wells
- Centering of the spheroid in each well enables automated visualization
- Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
- Ease of media exchange or treatment with test compounds/drugs
Legal Information
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Produkty
Discover how Corning® Spheroid Microplates played a key role in the use of use of 3D cell cultures for in vitro drug discovery assay.