Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
50 ea | Przewidywany termin wysyłki25 listopada 2026zSchnelldorf Distribution | 31 560,00 zł |
Informacje o tej pozycji
31 560,00 zł
Przewidywany termin wysyłki25 listopada 2026Szczegóły
material
polystyrene , round black/clear bottom
sterility
sterile
feature
manufacturer/tradename
Corning 4527
size
1536 wells
well volume
(14 mL)
wells
(1536)
suitability
suitable for (cell cuture)
General description
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel proprietary well geometry aids formation of spheroids in center of well
- Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
- Uniform, single spheroid formation across all wells
- Centering of the spheroid in each well enables automated visualization
- Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
- Ease of media exchange or treatment with test compounds/drugs
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| size 1536 wells | size 384 wells | size 96 wells | size 384 wells |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| material polystyrene , round black/clear bottom | material clear bottom, polystyrene , round black/clear bottom | material clear bottom, polystyrene , round black/clear bottom | material clear bottom, polystyrene , round black/clear bottom |
| feature | feature lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface) | feature lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface) | feature lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface) |
| manufacturer/tradename Corning 4527 | manufacturer/tradename Corning 3830 | manufacturer/tradename Corning 4520 | manufacturer/tradename Corning 4516 |
| wells (1536) | wells 384 wells | wells 96 wells | wells 384 wells |
Still not finding the right product?
Explore all of our products under Corning® spheroid microplates
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?