This plunger is 304 stainless steel coated with PTFE. Yes, there is a PTFE plunger tip on this syringe to help create the gas-tight seal.
Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 457,00 zł |
Informacje o tej pozycji
457,00 zł
material
blunt tip stainless steel needle (point style 3), glass body
needle size
22 ga (blunt tip)
feature
blunt tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81165
needle L
51 mm (2 in.)
volume
250 μL
application(s)
general analytical
compatibility
for use with Rheodyne, for use with SSI, for use with Valco VISF-2
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 1725RNR, volume 250 μL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.) | description 1705RNR, volume 50 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.) | description 1702RNR, volume 25 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.) | description 7001, volume 1 μL, needle size 25s ga (blunt tip), needle L 70 mm (2.75 in.) |
| material blunt tip stainless steel needle (point style 3), glass body | material blunt tip stainless steel needle (point style 3), glass body | material blunt tip stainless steel needle (point style 3), glass body | material blunt tip stainless steel needle (point style 3) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| needle size 22 ga (blunt tip) | needle size 22s ga (blunt tip) | needle size 22s ga (blunt tip) | needle size 25s ga (blunt tip) |
| manufacturer/tradename Hamilton 81165 | manufacturer/tradename Hamilton 80965 | manufacturer/tradename Hamilton 80265 | manufacturer/tradename Hamilton 80107 |
| feature blunt tip | feature blunt tip | feature blunt tip | feature KH (Knurled Hub) |
| compatibility for use with Valco VISF-2, for use with Rheodyne, for use with SSI | compatibility for use with SSI, for use with Valco VISF-2, for use with Rheodyne | compatibility for use with Valco VISF-2, for use with SSI, for use with Rheodyne | compatibility - |
Still not finding the right product?
Explore all of our products under Hamilton® HPLC syringes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
-
Which material is the plunger made of? Does it have a rubber end like many Hamilton syringes?