Kluczowe dokumenty
TANKVNT22
Milli-Q
Vent Filter
pore size 0.22 μm, carbon, and soda lime, Advanced protection against airborne contaminants for storage tanks
Wybierz wielkość
2 PKG
4930,00 zł
4930,00 zł
Przewidywany termin wysyłki15 kwietnia 2025Szczegóły
Wybierz wielkość
Zmień widok
2 PKG
4930,00 zł
About This Item
Kod UNSPSC:
41104212
NACRES:
NB.24
4930,00 zł
Przewidywany termin wysyłki15 kwietnia 2025Szczegóły
Poziom jakości
zastosowanie
(for type 3 water)
opakowanie
pkg of 2 units
producent / nazwa handlowa
Milli-Q®
Wysokość
33 cm (12.8 in.)
wielkość porów
0.22 μm pore size
kompatybilność
for use with SDS
for use with tanks
Powiązane kategorie
Opis ogólny
  • The vent filter protects highly sensitive stored pure water from possible contaminants in the environment. These include carbon dioxide, particles, micro-organisms and volatile organic compounds that mainly come from the laboratory atmosphere.
  • Contain a combination of purification media carefully selected to remove airborne contaminants that could degrade purified water quality during storage. Contains soda lime to prevent CO2 contamination.
  • The vent filter cartridges are constructed from polypropylene, polyethersulfone and styrene-acrylonitrile resin.
Zastosowanie
Vent filter provides advanced protection against airborne contaminants.
Cechy i korzyści
Soda Lime for carbon dioxide removal.
Informacje prawne
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany