Kluczowe dokumenty
SQPAKACARE
Milli-Q
SQPAK Care A Cleaner
Anti-scaling agent for use with Milli-Q® SQ 2Series RO production module.
About This Item
Kod UNSPSC:
41104212
eCl@ss:
33050190
NACRES:
NB.35
Produkt SQPAKACARE nie jest obecnie dostępny w sprzedaży w Twoim kraju Skontaktuj się z zespołem ds. pomocy technicznej
Poziom jakości
opakowanie
pkg of 1 unit
kompatybilność
for use with Milli-Q® SQ 2Series
Opis ogólny
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The SQPAK Care A Cleaner prevents scaling to safeguard the pure RO water production station of Milli-Q® SQ 2Series water purification systems.
The purification technology of the SQPAK Care A Cleaner uses an acidic reagent to remove mineral scaling in hydraulic parts. This enhances system performance and extends its lifetime.
The Twist and Lock mechanism of the cartridge enables easy replacement.
For use exclusively on the pure RO water production station of Milli-Q® SQ 2Series configurations, including Milli-Q® SR 240/240L/240XL and Milli-Q® SQ 240C/240CV systems.
Informacje prawne
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany