Kluczowe dokumenty
Z253014
Sigma-Aldrich
Enviro-Safe® thermometer
greener alternative
temperature -10-225 °C, immersion level, total
About This Item
Kod UNSPSC:
41112303
NACRES:
NB.79
Właściwości
immersion level total
producent / nazwa handlowa
HB Instruments 20520
charakterystyka ekologicznej alternatywy
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
zakres pomiaru temperatury
-10-225 °C
Długość
355 mm
kategoria ekologicznej alternatywy
Opis ogólny
Enviro-Safe thermometers are serialized, calibrated with NIST (NBS) traceable standards, and include a Statement of Accuracy.
White glass with easy-to-read, non-hazardous, biodegradable, green liquid fill. 6.5 mm O.D. guaranteed accuracy: ±1 scale division below 105°C (221°F), ±1.5 scale divisions above 105°C (221°F), and ±2 scale divisions above 200°C (392 °F).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemicals without mercury.
Informacje prawne
Enviro-Safe is a registered trademark of H-B Instrument Co.
produkt powiązany
Numer produktu
Opis
Cennik
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)