Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 425,00 zł |
Informacje o tej pozycji
425,00 zł
material
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
agency
suitable for DIN 38407-42
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3119-0010
nominal capacity
10 mL
General description
Features and Benefits
- Provides superior chemical resistance compared to polycarbonate tubes.
- Autoclavable and can withstand repeated autoclaving, resulting in a longer lifespan than polycarbonate tubes subjected to the same conditions.
- Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
- Leakproof when used with the provided Nalgene closure.
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description nominal capacity 10 mL | description nominal capacity 30 mL | description nominal capacity 28 mL | description nominal capacity 30 mL |
| sterility non-sterile | sterility non-sterile | sterility - | sterility non-sterile |
| manufacturer/tradename Nalgene® 3119-0010 | manufacturer/tradename Nalgene® 3119-0030 | manufacturer/tradename Nalgene® 3119-0028 | manufacturer/tradename Nalgene® 3118-0030 |
| material polypropylene copolymer , round bottom tube, polypropylene screw closure, screw top cap (Linerless) | material polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless) | material polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless) | material polypropylene screw closure, round bottom tube, screw top cap, transparent polycarbonate |
| feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea |
| agency suitable for DIN 38407-42 | agency - | agency - | agency - |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge tubes, Oak Ridge Style 3119
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?