Z113530
Electrode connector adapter
BNC meter to US Standard electrode
About This Item
Kod UNSPSC:
41115603
NACRES:
NB.79
Polecane produkty
Opis ogólny
If the type of connector on your pH meter is not compatible with your electrode, choose an appropriate adapter.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)