Key Documents
S9027
载玻片
opaque (white), size 25 mm × 75 mm
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122600
NACRES:
MA.01
材料
glass
包裝
case of 20
box of 72 slides
尺寸
25 mm × 75 mm
結合類型
non-treated surface
正在寻找类似产品? 访问 产品对比指南
一般說明
密封在吸湿干燥剂包装,同样可用作干燥架和储存盒。载玻片1mm厚。
我们的显微镜载玻片是由高质量白玻璃制成,含铁量低,没有淡绿色,质地良好,透明。这些载玻片还防腐蚀。边缘光滑,没有尖利的切割表面,使用安全。它们装在特殊的热成型塑料盒,能够减少和湿气灰尘接触(每个载玻片都预清理过)。
符合联邦规范NNN-S-450。
访问文档库
其他客户在看
Slide 1 of 1
1 of 1
盖玻片 size 22 mm × 22 mm
C9802
盖玻片
Xiao Yan et al.
ACS nano, 14(10), 12796-12809 (2020-10-15)
Droplet transport on, and shedding from, surfaces is ubiquitous in nature and is a key phenomenon governing applications including biofluidics, self-cleaning, anti-icing, water harvesting, and electronics thermal management. Conventional methods to achieve spontaneous droplet shedding enabled by surface-droplet interactions suffer
相关内容
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.