主要文件
DWKWCL0350-1
WHEATON® CELLine Multiuse Bioreactor flasks for suspension cell culture
polystyrene, flask capacity (350 mL), case of 1 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122104
NACRES:
NB.13
物料
molded polypropylene neck
polystyrene
screw top plastic cap
无菌性
sterile; γ-irradiated
特点
with cap
包装
case of 1 ea
制造商/商品名称
WHEATON® WCL0350-1
培养瓶容量
(350 mL)
正在寻找类似产品? 访问 产品对比指南
一般描述
CELLine Bioreactor Flask for high density suspension cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
  • Disposable and ready-to-use
  • High call density and high product concentration
  • Bulk media chamber allows for longer run times without media refreshing and reduces operation time
  • Decreases use of consumables
  • Cost-efficient, space saving, and stackable
  • No additional equipment required for operation
特点和优势
  • Ideal for high density cell culture, designed to enhance small scale bio-production for antibody and protein generation
  • Available in 1000ml flask for suspension (CELLine 1000) or adherent (CELLine 1000-AD) cell culture and 350ml flask for suspension culture only
  • Disposable and ready-to-use
  • Multi-harvest operation ensures a cost efficient solution for antibody and protein production
  • Ideal for use with cell lines such as Hybridoma, CHO, HEK, BHK, Sf9
法律信息
WHEATON is a registered trademark of Wheaton Industries, Inc.
历史批次信息供参考:
分析证书(COA)