Sprengstoffe in Wasser
Materialien
SPME-Faser
Produkt-Nr.
Beschreibung
Preisangaben
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB)
65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hubCONDITIONS
sample/matrix
50 ng/mL each explosive in water
SPME fiber
polydimethylsiloxane/divinylbenzene, 65 μm (57310-U)
extraction
immersion, 30 min
desorption process
5 min, 250 °C
column
14% cyanopropylphenyl polysiloxane, 30 m × 0.25 mm I.D., 0.25 μm (Supelco equivalent, Equity-1701, 28372-U)
oven
95 °C (3 min) to 182 °C at 8 °C/min (4 min) to 250 °C at 8 °C/min (6 min)
inj. temp.
split/splitless, 180 °C
detector
ECD, 250 °C
carrier gas
helium