57195U
57195-U
₩112,056
1 of 10
Supelco
575650-U
PlatePrep 96-well Vacuum Manifold
57030-U
Visiprep™ SPE Vacuum Manifold, PTFE Free
57265
Visiprep™ SPE Vacuum Manifold
57059
Disposable Liners for Visiprep DL Manifolds (included with 57044 and 57265)
57044
53997-U
Ascentis® Express C8 Capillary, 2.7 μm HPLC Column
57192-U
57029
Visiprep™ Manifold Cover (flow control valves and gasket included)
57161-U
SPE Manifold Gauge/Bleed Valve
Millipore
VPMG201NB1
Viresolve® Pro Device
solid phase extraction (SPE): suitable
food and beverages
for use with Supelco Plateprep Vacuum Manifold
Analytical Sample Preparation
Solid Phase Extraction (SPE) and QuEChERS
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.
도움이 필요하시면 연락하세요. 고객 지원 부서
문서 라이브러리 방문
1 of 5
Sigma-Aldrich
VM01
GenElute™ 96 Vacuum Manifold
575653-U
SPE 96-Deep Square Well Collection Plate
57250-U