58643
₩144,841
1 of 10
HAM80075
Hamilton® syringe
20890-U
Hamilton® HPLC syringes
26284
Hamilton® SampleLock syringe
20997
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
20888
22192-U
Hamilton® syringe, 1000 series GASTIGHT®, removable needle
HAM7657-01
Hamilton® TLC syringe
28637-U
Hamilton® 1001LTN syringe
20889
20734
Hamilton® syringe, 700 series, fixed needle
blunt tip stainless steel needle (point style 3)
22 ga (blunt tip)
Hamilton 81365
51 mm
1 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
Analytical Chromatography
Analytical Syringes
Hamilton Syringes
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.
도움이 필요하시면 연락하세요. 고객 지원 부서
문서 라이브러리 방문
1 of 9
28621-U
58383
Hamilton® syringe, 700 series, special needle
20886
Sigma-Aldrich
20784
Hamilton® GASTIGHT® syringe, 1700 series, removable needle
Z118109
Hamilton® needles for RN syringes
19139-U
Hamilton® needles for RN syringes, blunt tip
HAM204052
Hamilton® chromatography syringe