주요 문서
Z569933
Sigma-Aldrich
Chem-FLEX transfer line with male and female Luer connectors
tube L 60 in.
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩214,368
₩214,368
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
1 EA
₩214,368
About This Item
UNSPSC 코드:
41121815
NACRES:
NB.35
₩214,368
구입 가능 여부는 고객센터에 문의하십시오.
튜브 길이
60 in.
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
One male and one female Luer lock connector with chrome-plated brass bodies are connected to Chem-Flex 106 tubing with nylon clamps. Chem-Flex tubing was developed by Aldrich for the safe transfer of aggressive solvents, air-sensitive liquids, and gases.
Chem-Flex Tubing Features:
Chemically inert, thin-wall PTFE inner tube bonded to a clear PVC outer tube for extra strong, flexible transfer lines that are resistant to crushing or kinking.
  • Luer fittings on ends of tubing allow easy connection to
  • Stopcocks and valves
  • Needles of all lengths and gauges
  • Syringes and pumps
Applications:
  • Pressure transfer of liquids- Using Luer needles, stopcocks
  • Reagent addition to reactions- Using Luer needle, stopcock, syringe or syringe pump
법적 정보
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)