Z220329
Z220329-1EA
₩122,511
1 of 10
Sigma-Aldrich
Z166898
Aldrich® air-sensitive chemistry glass lab kit
Z516082
Schlenk reaction and storage tube
Z230855
Aldrich® Schlenk-type male joint cap
Z167533
Vacuum adapter
Z173061
Solvent storage flask with glass stopcock on side-arm
Z174300
Single-bulb ball-tube
Z173126
Reaction and storage tube with glass stopcock on side-arm
W389755
2-Propanethiol
Z167525
Aldrich® Schlenk-type Liebig condenser
Z173096
JK
female joint: ST/NS45/40
유사한 제품을 찾으십니까? 방문 제품 비교 안내
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.
도움이 필요하시면 연락하세요. 고객 지원 부서
문서 라이브러리 방문
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.