Z220264
Z220264-1EA
₩227,969
1 of 10
SYNF891410
Synthware™ reaction tube with glass stopcock
Sigma-Aldrich
Z166898
Aldrich® air-sensitive chemistry glass lab kit
Z173126
Reaction and storage tube with glass stopcock on side-arm
Z515981
Schlenk reaction and storage tube
Z409235
Aldrich® Schlaker reaction tube
Z208221
Rubbermaid® heavy-duty lab carts
SYNV165045
Synthware™ vacuum trap with serrated hose connections
Z173061
Solvent storage flask with glass stopcock on side-arm
SYNM181004
Synthware™ all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
SYNF531100
Synthware™ single neck reaction flask with glass stopcock
pear-shaped single-neck stainless steel
Q
male joint: ST/NS14/20
non-sterile
closure type Standard Taper Joint
pack of 1
25 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
Glassware
Laboratory Containers & Storage
Laboratory Flasks
Sample Handling
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.
도움이 필요하시면 연락하세요. 고객 지원 부서
문서 라이브러리 방문
1 of 7
Z173223
Z220434
Sealing tube
Z173363
Recrystallizer
Z173495
Addition funnel with Rotaflo® stopcock, and glass stopcock on side-arm
Z167525
Aldrich® Schlenk-type Liebig condenser
Z516007
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.