주요 문서
Z113255
Sigma-Aldrich
Sigma-Aldrich® standard Ag/AgCl double junction pH combination electrode, glass body
Standard pin, electrode L 55 mm , with bulb guard
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩390,572
₩390,572
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
1 EA
₩390,572
About This Item
UNSPSC 코드:
41115603
NACRES:
NB.79
₩390,572
구입 가능 여부는 고객센터에 문의하십시오.
전극 길이
55 mm , with bulb guard
스템 O.D.
8 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The combination of a pH indicating and Ag/AgCl reference half-cell in one electrode provides ease of handling and smaller sampling requirements. Combination double junction electrodes are Tris compatible with a maximum temperature of 80°C, and are recommended for proteins, metal ions, sulfides or other substances that will react with either Ag or AgCl. Combination Ag/AgCl electrodes are suggested for higher temperature applications (100°C). Both types supplied with a 30 in. lead.
Temperature Range:
  • -5 to 100°C Continuous Use
  • -5 to 130°C Intermittent Use
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
특징 및 장점
Short, narrow stem with bulb guard
법적 정보
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)