주요 문서
DWK92812F-20
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (20 mL), class A
로그인조직 및 계약 가격 보기
크기 선택
6 EA
₩481,212
₩481,212
예상 입고일2025년 3월 10일세부사항
크기 선택
보기 변경
6 EA
₩481,212
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
₩481,212
예상 입고일2025년 3월 10일세부사항
material
PTFE stopper
glass flask
특징
class A
포장
case of 6 ea
제조업체/상표
Kimble® 92812F-20
플라스크 용량
(20 mL)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
특징 및 장점
• Manufactured from borosilicate glass 3.3• Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets• Heavy duty walls provide superior strength, durability and safety• Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills• Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets• Printed large volume numbers allow quick identification when selecting glassware• Flat interior bottoms for stirring with standard stir bars• Calibrate to contain• Graduated to Class A volumetric tolerances•
법적 정보
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
Lot/Batch Number
죄송합니다. 지금은 이 제품에 대한 COA이(가) 온라인에서 제공되지 않습니다.
문서 라이브러리 방문
질문
  1. Can a 1-mL pipette fit into the neck of this volumetric flask?
    1 답변
    1. Yes a 1-mL pipette can fit into the neck of this volumetric flask.