주요 문서
CAD7974
Micro-emulsifying needle
with reinforcing bar, gauge 25 ×, L 2 7/8 in.
로그인조직 및 계약 가격 보기
크기 선택
4 EA
₩888,836
₩888,836
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
4 EA
₩888,836
About This Item
UNSPSC 코드:
42142546
NACRES:
NB.03
₩888,836
구입 가능 여부는 고객센터에 문의하십시오.
포장
pack of 4 ea
제조업체/상표
Cadence Science 7974
길이
2 7/8 in.
게이지
25 ×
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Widely used for allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity. By locking two syringes on to the double hub needle or double hub adapter, two liquids such as a mineral oil and an emulsifying agent may be readily pumped between the two syringes to produce a uniform mixture.
Indicated lengths are the overall length. Researchers and technicians utilize Micro-Mate® 5 mL and 10 mL Luer lock tip glass syringes for micro-emulsifying procedures. The outstanding quality of the glass and the special secure Luer locking tip provide quality and reliability.
법적 정보
Micro-Mate is a registered trademark of Cadence Science
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)