HPLCによるN-デスメチルネホパムエナンチオマーの分析、 CHIROBIOTIC® V2
材料
分析カラム
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] ammonium hydroxide; (100:0.1:0.02, A:B:C)
flow rate
1.2 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
2 mg/mL in methanol