HPLCによるミアンセリンエナンチオマーの分析、 CHIROBIOTIC® V
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (99.8:0.1:0.1, A:B:C)
flow rate
1 mL/min
column temp.
29 °C
detector
UV, 254 nm