HPLCによるメタドンエナンチオマーの分析、Astec® CHIROBIOTIC® V2
![HPLCによるメタドンエナンチオマーの分析、Astec® CHIROBIOTIC® V2 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/249/318/2f386abd-2ba7-47c2-8e55-763739c3eed0/800/2f386abd-2ba7-47c2-8e55-763739c3eed0.jpg)
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] 20 mM ammonium formate; (95:5, A:B)
flow rate
1 mL/min
column temp.
20 °C
detector
UV, 205 nm
injection
2 μL
sample
methadone, 1 mg/mL in mobile phase