Merck
GCによるピーナツバター中のピラジン分析、 SUPELCOWAX 10、SPME処理、50/30 μm DVB/Carboxen/PDMS使用
GCによるピーナツバター中のピラジン分析、 SUPELCOWAX 10、SPME処理、50/30 μm DVB/Carboxen/PDMS使用 suitable for GC, application for SPME
材料
SPMEファイバー
製品番号
詳細
価格
SPMEファイバーアセンブリー ジビニルベンゼン/カルボキセン/ポリジメチルシロキサン (DVB/CAR/PDMS)
50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Standard
製品番号
詳細
価格
二硫化炭素
anhydrous, ≥99%
2-エチルピラジン
≥98%, FG
2-メチルピラジン
Pharmaceutical Secondary Standard; Certified Reference Material
ジメチルジスルフィド
analytical standard
2,3-ジエチルピラジン
reference material
2,5-ジメチルピラジン
analytical standard
分析カラム
製品番号
詳細
価格
SUPELCOWAX 10キャピラリーGCカラム
L × I.D. 30 m × 0.25 mm, df 0.25 μm
関連製品
製品番号
詳細
価格
SPMEファイバーホルダー
for use with Manual Sampling, pk of 1
SPMEファイバーホルダー
for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1
CONDITIONS
sample/matrix
5 g peanut butter in 40 mL vial
SPME fiber
divinylbenzene-Carboxen-polydimethylbenzene StableFlex (57328-U)
extraction
headspace, 30 min at 65 °C in heating block
desorption process
270 °C for 5 min
column
SUPELCOWAX 10, 30 m × 0.25 mm, 0.25 μm (24079)
oven
40 °C (5 min) to 230 °C at 4 °C/min
inj. temp.
270 °C
injection
splitless/split, closed 0.5 min, 270 °C, with 0.75 mm liner
detector
ion trap mass spectrometer, m/z =30-350 at 0.6 sec/scan. Selected ions used for quantitation.
carrier gas
Helium, 30 cm/sec
liner
0.75 mm
sample
peanut butter
詳細
アナリシスノート
In this application, SPME was used for the analysis of volatiles in multifloral honey using a headspace method. Headspace SPME (HS-SPME) using a Carboxen/PDMS/DVB fiber coupled with gas chromatography-mass spectrometry (GC/MS) was used for the characterization of the volatile fraction of honey samples. Using SPME it was possible to obtain and distinguish the chemical fingerprints of different types of honey. The relatively polar SUPELCOWAX 10 GC column provides excellent separation of these polar compounds.